Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Methoxy-5-methylphenyl)-3-phenylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. M23675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23675G Supplier TCI America Supplier No. M23675G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109089-77-2
Molecular Formula C17H18O3
Molecular Weight (g/mol) 270.328
MDL Number MFCD01098026
InChI Key KSLPLDLDQANKOD-UHFFFAOYSA-N
PubChem CID 3468322
IUPAC Name 3-(2-methoxy-5-methylphenyl)-3-phenylpropanoic acid
SMILES CC1=CC(=C(C=C1)OC)C(CC(=O)O)C2=CC=CC=C2

Specifications

Melting Point 138°C
Color White
Quantity 5 g
Formula Weight 270.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Methoxy-5-methylphenyl)-3-phenylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.