Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(2-methoxyphenyl)-1-(2-pyridin-3-ylquinazolin-4-yl)urea | 280570-45-8 | 99.0% | 371.39 g/mol | C21H17N5O2 | 10 MG
SDP

Catalog No. 5000247450
Encompass_Preferred

VUF-5574 is a potent, selective antagonist of the adenosine A3 receptor used in pharmacology research to probe A3 receptor binding and signaling. It exhibits high affinity for the human recombinant A3 receptor and is supplied as a high-purity research compound.

  • Selective antagonist of adenosine A3 receptor (Ki = 4.03 nM)
  • High purity (99.0%)
  • Chemical formula C21H17N5O2; molecular weight 371.39 g/mol
  • Soluble in DMSO (≈100 mg/mL); formulatable for in vivo (10% DMSO/90% corn oil)
  • Recommended storage: powder -20°C; in solvent -80°C for long-term
  • Supplied in small research pack sizes suitable for in vitro and in vivo studies

Catalog No. 50-002-47450 Supplier Medchemexpress LLC Supplier No. HY10318910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VUF-5574 is a potent, selective antagonist of the adenosine A3 receptor used in pharmacology research to probe A3 receptor binding and signaling. It exhibits high affinity for the human recombinant A3 receptor and is supplied as a high-purity research compound.

  • Selective antagonist of adenosine A3 receptor (Ki = 4.03 nM)
  • High purity (99.0%)
  • Chemical formula C21H17N5O2; molecular weight 371.39 g/mol
  • Soluble in DMSO (≈100 mg/mL); formulatable for in vivo (10% DMSO/90% corn oil)
  • Recommended storage: powder -20°C; in solvent -80°C for long-term
  • Supplied in small research pack sizes suitable for in vitro and in vivo studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.