Learn More
Medchemexpress LLC 3-(2-methyl-1-oxopropyl)-1-[2-oxo-3-(4-phenoxyphenoxy)propyl]-1H-indole-5-carboxylic acid | 1233706-88-1 | MFCD18206878 | >98.0% | 471.5 | C28H25NO6 | 25 MG
SDP
Catalog No. 5000239672
Encompass_Preferred
CAY10650 is a potent and selective cytosolic phospholipase A2α (cPLA2α) inhibitor (IC50 = 12 nM) used to study inflammatory signaling and the synthesis of arachidonic acid-derived mediators. The compound is characterized by formula C28H25NO6, molecular weight 471.5, and is supplied at high purity for research use.
  • Inhibits cPLA2α with low nanomolar potency (IC50 = 12 nM).
  • Suppresses lipid droplet formation and PGE2 secretion in cellular assays.
  • Shows topical anti-inflammatory effects in mouse models.
  • Provided at ≥98% purity for research applications.
  • Suitable for studies of prostaglandin and leukotriene biosynthesis.
Catalog No. 50-002-39672 Supplier Medchemexpress LLC Supplier No. HY1080125MG
CAY10650 is a potent and selective cytosolic phospholipase A2α (cPLA2α) inhibitor (IC50 = 12 nM) used to study inflammatory signaling and the synthesis of arachidonic acid-derived mediators. The compound is characterized by formula C28H25NO6, molecular weight 471.5, and is supplied at high purity for research use.
  • Inhibits cPLA2α with low nanomolar potency (IC50 = 12 nM).
  • Suppresses lipid droplet formation and PGE2 secretion in cellular assays.
  • Shows topical anti-inflammatory effects in mouse models.
  • Provided at ≥98% purity for research applications.
  • Suitable for studies of prostaglandin and leukotriene biosynthesis.