Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[(2-methylphenyl)methylsulfanyl]-6-pyridin-2-ylpyridazine | 894002-50-7 | ≥98.0% | 293.39 g·mol-1 | C17H15N3S | 5 MG
SDP

Catalog No. 5000264345
Encompass_Preferred

LDN-212320 is a small-molecule activator of the glutamate transporter GLT-1 (EAAT2) used in research to increase EAAT2 expression and probe glutamate transport-related biology, including pain models. It is supplied as a powder and in solution formats suitable for in vitro and in vivo studies.

  • Activates GLT-1/EAAT2 at the translational level.
  • Small molecule; chemical formula C17H15N3S; molecular weight 293.39 g·mol-1.
  • High purity (≥98.0% by HPLC).
  • Available in mg-scale vials and as a 10 mM solution in DMSO for stock preparation.
  • Soluble in DMSO (50 mg/mL); in vivo formulations achievable at ≥2.5 mg/mL with appropriate co-solvents.
  • Storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C recommended.
  • Used in preclinical studies to modulate glutamate uptake and attenuate nociceptive and chronic pain phenotypes.

Catalog No. 50-002-64345 Supplier Medchemexpress LLC Supplier No. HY127415MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LDN-212320 is a small-molecule activator of the glutamate transporter GLT-1 (EAAT2) used in research to increase EAAT2 expression and probe glutamate transport-related biology, including pain models. It is supplied as a powder and in solution formats suitable for in vitro and in vivo studies.

  • Activates GLT-1/EAAT2 at the translational level.
  • Small molecule; chemical formula C17H15N3S; molecular weight 293.39 g·mol-1.
  • High purity (≥98.0% by HPLC).
  • Available in mg-scale vials and as a 10 mM solution in DMSO for stock preparation.
  • Soluble in DMSO (50 mg/mL); in vivo formulations achievable at ≥2.5 mg/mL with appropriate co-solvents.
  • Storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C recommended.
  • Used in preclinical studies to modulate glutamate uptake and attenuate nociceptive and chronic pain phenotypes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.