Learn More
Beckman Coulter 3.2 ML POYLMER TUBBING PK/50
Catalog No. NC9165843
Encompass_Preferred
Thickwall Polyallomer Tubes, Capacity: 3.2mL, Nominal Size: 13 x 56mm, For TLA-110, TLA-100.4 Rotors
Catalog No. NC9165843 Supplier Beckman Coulter Supplier No. 362333
Thickwall Polyallomer Tubes, Capacity: 3.2mL, Nominal Size: 13 x 56mm, For TLA-110, TLA-100.4 Rotors