Learn More
BioSpec Products Stainless Steel Beads 3.2mm 90g
SDP
Catalog No. NC0445832
Encompass_Preferred
Stainless steel beads have a density of 7.9g/cc. They are used mostly for grinding leaves and seeds. They are made of Type 316 stainless steel - the most corrosion resistant available.
Catalog No. NC0445832 Supplier BioSpec Products Supplier No. 11079132SS
Stainless steel beads have a density of 7.9g/cc. They are used mostly for grinding leaves and seeds. They are made of Type 316 stainless steel - the most corrosion resistant available.