Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[3-(2-Morpholinoethoxy)phenyl]methanol, 97%, Thermo Scientific™

Catalog No. CC42009DA
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC42009DA 1 g
CC42009CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC42009DA Supplier Thermo Scientific Chemicals Supplier No. CC42009DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 857284-07-2
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD07772846
InChI Key DZHYOLUKMGNRTO-UHFFFAOYSA-N
Synonym 3-2-morpholinoethoxy phenyl methanol, 3-2-morpholin-4-yl ethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methan-1-ol
PubChem CID 7164622
IUPAC Name [3-(2-morpholin-4-ylethoxy)phenyl]methanol
SMILES C1COCCN1CCOC2=CC=CC(=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material [3-(2-Morpholinoethoxy)phenyl]methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.