Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Naphthyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog No. AAH5294803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5294803 1 g
AAH5294806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5294803 Supplier Thermo Scientific Chemicals Supplier No. H5294803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870774-29-1
Molecular Formula C16H13BO2
Molecular Weight (g/mol) 248.09
MDL Number MFCD16293738
InChI Key WLKQDOGZCIYEOM-UHFFFAOYSA-N
Synonym 3-2-naphthyl phenylboronic acid, 3-naphthalen-2-yl phenyl boronic acid, 3-2-naphthyl benzeneboronic acid, 3-naphthalene-2-yl phenylboronic acid, boronic acid, 3-2-naphthalenyl phenyl, 3-naphthalen-2-yl phenylboronic acid, 3-2-naphthyl phenylboronicacid, 3-2-naphthyl phenyl boronic acid
PubChem CID 45382255
IUPAC Name (3-naphthalen-2-ylphenyl)boronic acid
SMILES OB(O)C1=CC(=CC=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Quantity 1 g
Formula Weight 248.09
Percent Purity 98%
Chemical Name or Material 3-(2-Naphthyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.