Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. N10095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1009-5G 5 g
N10091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1009-5G Supplier TCI America Supplier No. N10095G
Add to Cart
Add to Cart

Specifications

CAS 870774-29-1
Color White-Yellow
Molecular Formula C16H13BO2
MDL Number MFCD16293738
Quantity 5 g
Synonym 3-2-naphthyl phenylboronic acid, 3-naphthalen-2-yl phenyl boronic acid, 3-2-naphthyl benzeneboronic acid, 3-naphthalene-2-yl phenylboronic acid, boronic acid, 3-2-naphthalenyl phenyl
InChI Key WLKQDOGZCIYEOM-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)C2=CC3=CC=CC=C3C=C2)(O)O
IUPAC Name (3-naphthalen-2-ylphenyl)boronic acid
Molecular Weight (g/mol) 248.088
PubChem CID 45382255
Formula Weight 248.09
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.