Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(2-Oxo-2-phenylethylthio)benzoic acid

Catalog No. OTV000483 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000483 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000483 Supplier Sigma Aldrich Supplier No. OTV000483250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,5-heptanedione has been used in:
  • Preparation of 3,5-diethylpyrazole hydrochloride
  • Enantioselective conjugate addition of 1,3-dicarbonyls to nitroolefins via nickel(ii)-diamine catalysis

Specifications

Molecular Formula C15H12O3S
Linear Formula C15H12O3S
MDL Number MFCD22025919
Quantity 5 g
Molecular Weight (g/mol) 272.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.