Learn More
Medchemexpress LLC 3-[(2-pyridinyl)methylene]indolin-2-one | 3367-88-2 | 99.1% | 222.24 | C14H10N2O | 10MG
SDP
Catalog No. 5000239575
Encompass_Preferred
(E/Z)-GSK-3β inhibitor 1 (CAS 3367-88-2) is a small-molecule research compound that inhibits glycogen synthase kinase-3 beta. Supplied as an orange to red solid, it has molecular formula C14H10N2O and a molecular weight of approximately 222.24 g·mol-1. The material is provided at high purity for biochemical and cell-based research and should be stored protected from light and handled as a research chemical.
  • Inhibits glycogen synthase kinase-3 beta in biochemical assays.
  • Supplied as an orange to red solid for laboratory use.
  • High purity suitable for research applications.
  • Available in small quantities appropriate for experimental workflows.
  • Store protected from light; in solvent store at -80°C for long term and -20°C for short term.
Catalog No. 50-002-39575 Supplier Medchemexpress LLC Supplier No. HY126144A10MG
Add to Cart
Add to Cart
(E/Z)-GSK-3β inhibitor 1 (CAS 3367-88-2) is a small-molecule research compound that inhibits glycogen synthase kinase-3 beta. Supplied as an orange to red solid, it has molecular formula C14H10N2O and a molecular weight of approximately 222.24 g·mol-1. The material is provided at high purity for biochemical and cell-based research and should be stored protected from light and handled as a research chemical.
  • Inhibits glycogen synthase kinase-3 beta in biochemical assays.
  • Supplied as an orange to red solid for laboratory use.
  • High purity suitable for research applications.
  • Available in small quantities appropriate for experimental workflows.
  • Store protected from light; in solvent store at -80°C for long term and -20°C for short term.