Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4,4-disulfonic acid sodium salt, ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. 111015276 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5276 5 g
11-101-5275 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5276 Supplier MilliporeSigma Supelco Supplier No. 829505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt also known as Ferrozine is a chelating agent for ferrous iron.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00078417
Synonym 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4 ′,4 ′′-disulfonic acid sodium salt; PDT disulfonate

Specifications

Melting Point ≥300°C
CAS 69898-45-9
Quantity 5 g
Linear Formula C20H13N4 NaO6S2
Formula Weight 492.46
Percent Purity ≥97.0% (T); ≥97.0%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.