Learn More
Medchemexpress LLC 3-(2-Pyridyldithio)propanoic acid | 68617-64-1 | 99.5% | 1 G
SDP
Catalog No. 5000338512
Encompass_Preferred
3-(2-Pyridyldithio)propanoic acid is an alkyl chain-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-38512 Supplier Medchemexpress LLC Supplier No. HY1301571G
3-(2-Pyridyldithio)propanoic acid is an alkyl chain-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only