Learn More
Medchemexpress LLC 3-(2-Pyridyldithio)propanoic Acid | 68617-64-1 | 99.5% | 25 G
SDP
Catalog No. 5000341978
Encompass_Preferred
3-(2-Pyridyldithio)propanoic Acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two ligands: one for an E3 ubiquitin ligase and another for the target protein.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41978 Supplier Medchemexpress LLC Supplier No. 25GHY130157
3-(2-Pyridyldithio)propanoic Acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two ligands: one for an E3 ubiquitin ligase and another for the target protein.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins