Learn More
Medchemexpress LLC 3-(2-Pyridyldithio)propanoic Acid | 68617-64-1 | 99.5% | 500 MG
SDP
Catalog No. 5000339938
Encompass_Preferred
3-(2-Pyridyldithio)propanoic Acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. It is intended for research use only and not sold to patients. This compound has a molecular weight of 215.29 and the formula C8H9NO2S2. It appears as a white to off-white solid.
  • Alkyl chain-based linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • White to off-white solid appearance
Catalog No. 50-003-39938 Supplier Medchemexpress LLC Supplier No. HY130157500MG
3-(2-Pyridyldithio)propanoic Acid is an alkyl chain-based PROTAC linker used in the synthesis of PROTACs. It is intended for research use only and not sold to patients. This compound has a molecular weight of 215.29 and the formula C8H9NO2S2. It appears as a white to off-white solid.
  • Alkyl chain-based linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • White to off-white solid appearance