Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Thienyl)-L-alanine 97.0+%, TCI America™
SDP

Catalog No. T3475200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3475200MG 200 mg
T34751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3475200MG Supplier TCI America Supplier No. T3475200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22951-96-8
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD00064335
InChI Key WTOFYLAWDLQMBZ-LURJTMIESA-N
Synonym 3-2-thienyl-l-alanine, s-2-amino-3-thiophen-2-yl propanoic acid, l-2-thienylalanine, beta 2-thienyl alanine, h-thi-oh, 3-thiophen-2-yl-l-alanine, 2s-2-amino-3-thiophen-2-ylpropanoic acid, tih, beta-2-thienylalanine, pubchem18715
PubChem CID 146719
IUPAC Name (2S)-2-amino-3-thiophen-2-ylpropanoic acid
SMILES C1=CSC(=C1)CC(C(=O)O)N

Specifications

Melting Point 235°C
Quantity 200 mg
Formula Weight 171.21
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(2-Thienyl)-L-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.