Learn More
Medchemexpress LLC 3α,21-dihydroxy-5α-pregnan-20-one | 567-02-2 | MFCD00051144 | 98.0% | 334.49 | C21H34O3 | 5MG
SDP
Catalog No. 5000237094
Encompass_Preferred
3α,21-Dihydroxy-5α-pregnan-20-one (THDOC) is an endogenous neurosteroid that acts as a positive allosteric modulator of GABAA receptors. It potentiates neuronal responses to low concentrations of GABA and is used in preclinical research on GABAergic signaling and neurological disorders.
  • Endogenous neurosteroid that modulates GABAA receptor activity.
  • Potentiates neuronal response at α4β1δ GABAA receptors in vitro.
  • Supplied at high purity (98.0%).
  • Available as solids (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and as a 10 mM solution in DMSO.
  • Intended for in vitro research on GABAergic signaling and neurological disease models.
Catalog No. 50-002-37094 Supplier Medchemexpress LLC Supplier No. HY1234895MG
3α,21-Dihydroxy-5α-pregnan-20-one (THDOC) is an endogenous neurosteroid that acts as a positive allosteric modulator of GABAA receptors. It potentiates neuronal responses to low concentrations of GABA and is used in preclinical research on GABAergic signaling and neurological disorders.
  • Endogenous neurosteroid that modulates GABAA receptor activity.
  • Potentiates neuronal response at α4β1δ GABAA receptors in vitro.
  • Supplied at high purity (98.0%).
  • Available as solids (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and as a 10 mM solution in DMSO.
  • Intended for in vitro research on GABAergic signaling and neurological disease models.