Learn More
Ace Glass, Inc. Combo mantle power control, 3 position, 240v 7.5amp output, percentage timer (pulse) type
SDP
Catalog No. 50977355
Encompass
3-position Power Control for multi-element combination heating mantle. Power control is the percentage timer (pulse) type. 240v 7.5amp output.
Catalog No. 50-977-355 Supplier Ace Glass, Inc. Supplier No. 1206163
3-position Power Control for multi-element combination heating mantle. Power control is the percentage timer (pulse) type. 240v 7.5amp output.