Learn More
Ace Glass, Inc. Combo mantle power control, 3 position, 240v 7.5amp output, proportional voltage (steady state) type
SDP
Catalog No. 50977358
Encompass
3-position Power Control for multi-element combination heating mantle. Power control is the proportional voltage (steady state) type. 240v 7.5amp output.
Catalog No. 50-977-358 Supplier Ace Glass, Inc. Supplier No. 1206173
3-position Power Control for multi-element combination heating mantle. Power control is the proportional voltage (steady state) type. 240v 7.5amp output.