Learn More
AdipoGen 3 28-didiMeGbetulin
SDP
Supplier:  AdipoGen BVT0404M005
Encompass_Preferred
Chemical. Formula C44H70O8. MW 727. Semi-synthetic derivative from betulin from birch bark. Semi-synthetic derivative of betulin from birch bark. Potent antiviral compound. Shows anti-HIV activity more active than AZT in vitro.
Catalog No. 50-256-1943