Learn More
TARGETMOL CHEMICALS INC 3 29-Dibenzoyl Rarounitriol
SDP
Catalog No. 502880119
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. 3,29-Dibenzoyl rarounitriol is the main active compound of the polychlorinated alkane triterpene esters in Trichosanthes kirilowii and can be used as a quantitative marker of Trichosanthes kirilowii. Purity 95.8%
Catalog No. 50-288-0119 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S102810MG
Also available in 1 mL and bulk. Please contact Fisher for quotes. 3,29-Dibenzoyl rarounitriol is the main active compound of the polychlorinated alkane triterpene esters in Trichosanthes kirilowii and can be used as a quantitative marker of Trichosanthes kirilowii. Purity 95.8%