Learn More
Medchemexpress LLC 3(2H)-Pyridazinone, 2,4-dimethyl-6-[1-[(1S)-1-phenylethyl]-6-(tetrahydropyran-4-yl)-1H-imidazo | 1660117-38-3 | 98.8% | 429.51 g/mol | C25H27N5O2 | 5 MG
SDP
Catalog No. 5000226625
Encompass_Preferred
BRD4 Inhibitor-10 is a potent small-molecule inhibitor selective for the BD1 bromodomain of BRD4 (reported IC50 8 nM). Provided as a 5 mg solid research reagent, its chemical identifiers include CAS 1660117-38-3, formula C25H27N5O2, and molecular weight 429.51 g/mol. Supplier-reported purity is >98%.
  • Potent BRD4-BD1 inhibition (IC50 8 nM).
  • High purity suitable for biochemical and cellular assays.
  • Well-defined small-molecule formula and molecular weight (C25H27N5O2; 429.51 g/mol).
  • Supplied as a solid (5 mg) with solution formats available for immediate use.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-26625 Supplier Medchemexpress LLC Supplier No. HY1174915MG
BRD4 Inhibitor-10 is a potent small-molecule inhibitor selective for the BD1 bromodomain of BRD4 (reported IC50 8 nM). Provided as a 5 mg solid research reagent, its chemical identifiers include CAS 1660117-38-3, formula C25H27N5O2, and molecular weight 429.51 g/mol. Supplier-reported purity is >98%.
  • Potent BRD4-BD1 inhibition (IC50 8 nM).
  • High purity suitable for biochemical and cellular assays.
  • Well-defined small-molecule formula and molecular weight (C25H27N5O2; 429.51 g/mol).
  • Supplied as a solid (5 mg) with solution formats available for immediate use.
  • Intended for research use only; not for human or clinical use.