Learn More
Medchemexpress LLC 3(2H)-Pyridazinone, 4,5-dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-d3 | 74150-27-9 | 99.4% | 100 MG
SDP
Catalog No. 5000407368
Encompass_Preferred
Pimobendan-d3 (UD-CG115-d3) is a deuterium-labeled analog of Pimobendan, a selective inhibitor of PDE3 with an IC50 of 0.32 μM. This isotope-labeled compound is valuable in drug development for quantitative analysis, serving as a tracer and an internal standard. Deuteration can also influence pharmacokinetic and metabolic profiles. Its high isotopic enrichment ensures reliability in analytical applications.
  • Can be used as a tracer.
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Isotopic enrichment of ≥99.99%.
  • Deuteration can potentially influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07368 Supplier Medchemexpress LLC Supplier No. HYB0204S1100MG
Pimobendan-d3 (UD-CG115-d3) is a deuterium-labeled analog of Pimobendan, a selective inhibitor of PDE3 with an IC50 of 0.32 μM. This isotope-labeled compound is valuable in drug development for quantitative analysis, serving as a tracer and an internal standard. Deuteration can also influence pharmacokinetic and metabolic profiles. Its high isotopic enrichment ensures reliability in analytical applications.
  • Can be used as a tracer.
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Isotopic enrichment of ≥99.99%.
  • Deuteration can potentially influence pharmacokinetic and metabolic profiles of drugs.