Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3(2H)-pyridazinone, 4,5-dihydro-6-[4-(4-pyridinylamino)phenyl]-, hydrochloride | 98326-33-1 | 99.8% | 302.76 g/mol | C15H15ClN4O | 1 ML
SDP

Catalog No. 5000247664
Encompass_Preferred

Senazodan hydrochloride is the monohydrochloride salt of Senazodan (MCI 154), a Ca2+ sensitiser that also exhibits inhibitory activity against phosphodiesterase III (PDE III). It is provided for research use in biochemical and pharmacological studies and is supplied in solid and solution formats for assay applications.

  • High purity (99.84%) as verified by certificate of analysis.
  • Exact chemical identity: C15H15ClN4O, MW 302.76 g/mol, CAS 98326-33-1.
  • Suitable for biochemical and pharmacological assays involving Ca2+ sensitization and PDE III modulation.
  • Available as solid powder and as concentrated solution formats for convenient dosing.
  • COA available for batch-specific quality and analytical data.

Catalog No. 50-002-47664 Supplier Medchemexpress LLC Supplier No. HY101693A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Senazodan hydrochloride is the monohydrochloride salt of Senazodan (MCI 154), a Ca2+ sensitiser that also exhibits inhibitory activity against phosphodiesterase III (PDE III). It is provided for research use in biochemical and pharmacological studies and is supplied in solid and solution formats for assay applications.

  • High purity (99.84%) as verified by certificate of analysis.
  • Exact chemical identity: C15H15ClN4O, MW 302.76 g/mol, CAS 98326-33-1.
  • Suitable for biochemical and pharmacological assays involving Ca2+ sensitization and PDE III modulation.
  • Available as solid powder and as concentrated solution formats for convenient dosing.
  • COA available for batch-specific quality and analytical data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.