Learn More
BioSpec Products Chrome-Steel Beads 3.2mm 1 lb
SDP
Catalog No. NC0778455
Encompass_Preferred
Chrome steel has a density of 7.9g/cc (used mostly for grinding leaves). Listed bead sizes are median (average) values.  Because beads are sorted by sieves, bead size varies between +/-10% of a listed value. 1 lb
Catalog No. NC0778455 Supplier BioSpec Products Supplier No. 11079132C
Chrome steel has a density of 7.9g/cc (used mostly for grinding leaves). Listed bead sizes are median (average) values.  Because beads are sorted by sieves, bead size varies between +/-10% of a listed value. 1 lb