Learn More
Cayman Chemical 3 3-2-Chlorophenylmethyle 10mg
SDP
Catalog No. 502988453
Encompass_Preferred
An ENPP1 inhibitor (Ki = 50 µM); inhibits urease (IC50 = 84.53 µM for the Jack bean enzyme)
Catalog No. 50-298-8453 Supplier Cayman Chemical Supplier No. 2986410 MG
An ENPP1 inhibitor (Ki = 50 µM); inhibits urease (IC50 = 84.53 µM for the Jack bean enzyme)