Learn More
Medchemexpress LLC 3-(3-(2-methoxyphenyl)-4-oxothiazolidin-2-yl)-N-(4-phenylbutyl)benzamide | 1415251-36-3 | 98.0% | 460.59 | C27H28N2O3S | 10MG
SDP
Catalog No. 5000237162
Encompass_Preferred
WJ460 is a small-molecule inhibitor of myoferlin (MYOF) used in research to explore tumor cell migration, invasion, and ferroptosis. It binds MYOF and has been reported to inhibit cell migration and growth, induce cell-cycle arrest, trigger mitochondrial autophagy, and promote lipid peroxidation. The compound is supplied as a purified research-grade solid for in vitro studies.
  • Potent myoferlin inhibitor with reported cellular activity.
  • Inhibits cell migration and proliferation in tumor models.
  • Induces cell-cycle arrest and mitochondrial autophagy.
  • Promotes lipid peroxidation and ferroptosis in cancer cells.
  • Provided as a purified solid suitable for in vitro assays.
  • Molecular weight and formula provided for experimental planning.
Catalog No. 50-002-37162 Supplier Medchemexpress LLC Supplier No. HY12463210MG
WJ460 is a small-molecule inhibitor of myoferlin (MYOF) used in research to explore tumor cell migration, invasion, and ferroptosis. It binds MYOF and has been reported to inhibit cell migration and growth, induce cell-cycle arrest, trigger mitochondrial autophagy, and promote lipid peroxidation. The compound is supplied as a purified research-grade solid for in vitro studies.
  • Potent myoferlin inhibitor with reported cellular activity.
  • Inhibits cell migration and proliferation in tumor models.
  • Induces cell-cycle arrest and mitochondrial autophagy.
  • Promotes lipid peroxidation and ferroptosis in cancer cells.
  • Provided as a purified solid suitable for in vitro assays.
  • Molecular weight and formula provided for experimental planning.