Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-(3,3'-Dimethoxy-4,4'-diphenylene)bis(2-phenyl-5-veratryltetrazolium Chloride), TCI America™
SDP

Catalog No. B0369100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0369100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0369100MG Supplier TCI America Supplier No. B0369100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106629-90-7
Molecular Formula C46H44Cl2N8O6
Molecular Weight (g/mol) 875.808
MDL Number MFCD00060036
InChI Key WYENYHSISIKTHQ-UHFFFAOYSA-L
PubChem CID 44630099
IUPAC Name 5-[(3,4-dimethoxyphenyl)methyl]-2-[4-[4-[5-[(3,4-dimethoxyphenyl)methyl]-3-phenyltetrazol-2-ium-2-yl]-3-methoxyphenyl]-2-methoxyphenyl]-3-phenyltetrazol-2-ium;dichloride
SMILES COC1=C(C=C(C=C1)CC2=NN([N+](=N2)C3=C(C=C(C=C3)C4=CC(=C(C=C4)[N+]5=NC(=NN5C6=CC=CC=C6)CC7=CC(=C(C=C7)OC)OC)OC)OC)C8=CC=CC=C8)OC.[Cl-].[Cl-]

Specifications

Quantity 100 mg
Formula Weight 875.81
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-(3,3′-Dimethoxy-4,4′-diphenylene)bis(2-phenyl-5-veratryltetrazolium Chloride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.