Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,3',3'-Tetramethyl-1,1'-bis(4-sulfobutyl)benzoindodicarbocyanine Sodium Salt 98.0+%, TCI America™
SDP

Catalog No. D44631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44631G 1 g
D44635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44631G Supplier TCI America Supplier No. D44631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64285-36-5
Molecular Formula C41H47N2NaO6S2+
Molecular Weight (g/mol) 750.945
MDL Number MFCD16621036
InChI Key ZOYVHUDSHDNKLZ-UHFFFAOYSA-O
Synonym Sodium 4-[2-[5-[1,1-Dimethyl-3-(4-sulfonatobutyl)-1H-benz[e]indol-2(3H)-ylidene]-1,3-pentadienyl]-1,1-dimethyl-1H-benz[e]indol-3-ium-3-yl]butane-1-sulfonate, 2-[5-[1,1-Dimethyl-3-(4-sulfobutyl)-1,3-dihydrobenzo[e]indol-2-ylidene]-1,3-pentadienyl]-1,1-dime
PubChem CID 129894317
IUPAC Name 4-[(2E)-2-[(2E,4E)-5-[1,1-dimethyl-3-(4-sulfobutyl)benzo[e]indol-3-ium-2-yl]penta-2,4-dienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonic acid;sodium
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)O)C=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)O)C=CC6=CC=CC=C65)(C)C)C.[Na]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 748.93
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,3′,3′-Tetramethyl-1,1′-bis(4-sulfobutyl)benzoindodicarbocyanine Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.