Learn More
Medchemexpress LLC 3,3,3-trideuterio-2-(3-fluoro-4-phenylphenyl)propanoic acid | 1185133-81-6 | MFCD09840635 | 99.9% | 247.28 g/mol | C15H10D3FO2 | 1 MG
SDP
Catalog No. 5000243272
Encompass_Preferred
Flurbiprofen-d3 is a deuterium-labeled analogue of flurbiprofen supplied as an isotopically labeled reference standard for analytical and research use. It is used primarily as an internal standard to enable accurate quantitation in mass spectrometry-based assays and bioanalytical method development.
  • Isotopically labeled internal standard for quantitative mass spectrometry.
  • High chemical and isotopic purity for reliable assay performance.
  • Suitable for LC-MS/MS and GC-MS bioanalytical workflows.
  • Applicable to pharmacokinetic and metabolic studies.
  • Provided as a stable solid with batch-specific documentation.
Catalog No. 50-002-43272 Supplier Medchemexpress LLC Supplier No. HY10582S1MG
Flurbiprofen-d3 is a deuterium-labeled analogue of flurbiprofen supplied as an isotopically labeled reference standard for analytical and research use. It is used primarily as an internal standard to enable accurate quantitation in mass spectrometry-based assays and bioanalytical method development.
  • Isotopically labeled internal standard for quantitative mass spectrometry.
  • High chemical and isotopic purity for reliable assay performance.
  • Suitable for LC-MS/MS and GC-MS bioanalytical workflows.
  • Applicable to pharmacokinetic and metabolic studies.
  • Provided as a stable solid with batch-specific documentation.