Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,3-Trifluoro-2,2-dimethylpropionic acid, Thermo Scientific™

Catalog No. MAY00264DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00264DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00264DA Supplier Thermo Scientific Chemicals Supplier No. MAY00264DA
Add to Cart

Chemical Identifiers

CAS 889940-13-0
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.104
InChI Key VMFSJVUPIXOCFO-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid
PubChem CID 19867367
IUPAC Name 3,3,3-trifluoro-2,2-dimethylpropanoic acid
SMILES CC(C)(C(=O)O)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material 3,3,3-trifluoro-2,2-dimethylpropionic acid
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.