Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,3-Trifluoropropionic acid, 98%

Catalog No. AAB2042814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2042814 25 g
AAB2042806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2042814 Supplier Thermo Scientific Chemicals Supplier No. B2042814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2516-99-6 | C3H3F3O2 | 128.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-99-6
Molecular Formula C3H3F3O2
Molecular Weight (g/mol) 128.05
MDL Number MFCD00153292
InChI Key KSNKQSPJFRQSEI-UHFFFAOYSA-N
Synonym 3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-propionic acid, propanoic acid, 3,3,3-trifluoro, tfpa, acmc-209ghu, trifluoromethylacetic acid, trifluoromethyl acetic acid, ksc203o6p, 3,3,3,-trifluoropropionic acid, 3,3,3-tri-fluoropropionic acid
PubChem CID 2777972
IUPAC Name 3,3,3-trifluoropropanoic acid
SMILES C(C(=O)O)C(F)(F)F

Specifications

Melting Point 9°C
Density 1.441
Boiling Point 146°C
Flash Point >100°C (212°F)
Refractive Index 1.334
Quantity 25 g
UN Number UN3265
Beilstein 1751796
Formula Weight 128.05
Percent Purity 98%
Chemical Name or Material 3,3,3-Trifluoropropionic acid
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P374-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.