Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3,3-trifluoropropylamine

Catalog No. CDS011723 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS011723 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS011723 Supplier Sigma Aldrich Supplier No. CDS011723100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dimethyl sulfite is an insect repellent and also experimentally identified as a mutagenic agent.the conformations and vibrational spectra of dimethyl sulfite was studied by matrix-isolation fourier transform infrared spectroscopy and to study its conformational selective aggregation
  • It may be used in organic synthesis

Specifications

CAS 460-39-9
Molecular Formula C3H6F3N
Linear Formula C3H6F3N
MDL Number MFCD01694854
Quantity 100 g
Molecular Weight (g/mol) 113.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.