Learn More
Medchemexpress LLC 3-(3-{4-[3-(β-D-glucopyranosyloxy)-5-isopropyl-1H-pyrazol-4-ylmethyl]-3-methylphenoxy}...) | 666842-36-0 | 98.8% | 536.62 | C26H40N4O8 | 50MG
SDP
Catalog No. 5000237666
Encompass_Preferred
KGA-2727 is a selective, high-affinity, orally active small-molecule inhibitor of the sodium-glucose cotransporter 1 (SGLT1) that has demonstrated antidiabetic efficacy in rodent models and inhibits SGLT1-mediated glucose uptake in a dose-dependent manner.
  • Selective SGLT1 inhibitor with Ki 97.4 nM (human) and 43.5 nM (rat).
  • High purity (98.8%).
  • Molecular formula C26H40N4O8 and molecular weight 536.62 g·mol⁻¹.
  • Available in multiple milligram pack sizes for research use.
  • Solid, white to yellow appearance; store solid at 4 °C and protect from light.
  • Includes supporting analytical documents such as certificate of analysis and safety data sheet.
Catalog No. 50-002-37666 Supplier Medchemexpress LLC Supplier No. HY12379750MG
KGA-2727 is a selective, high-affinity, orally active small-molecule inhibitor of the sodium-glucose cotransporter 1 (SGLT1) that has demonstrated antidiabetic efficacy in rodent models and inhibits SGLT1-mediated glucose uptake in a dose-dependent manner.
  • Selective SGLT1 inhibitor with Ki 97.4 nM (human) and 43.5 nM (rat).
  • High purity (98.8%).
  • Molecular formula C26H40N4O8 and molecular weight 536.62 g·mol⁻¹.
  • Available in multiple milligram pack sizes for research use.
  • Solid, white to yellow appearance; store solid at 4 °C and protect from light.
  • Includes supporting analytical documents such as certificate of analysis and safety data sheet.