Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(3-{4-[3-(β-D-glucopyranosyloxy)-5-isopropyl-1H-pyrazol-4-ylmethyl]-3-methylphenoxy}propylamino... | 666842-36-0 | 98.8% | 536.62 g/mol | C26H40N4O8 | 10MG
SDP

Catalog No. 5000236806
Encompass_Preferred

KGA-2727 is a selective, high-affinity, orally active SGLT1 inhibitor used in preclinical research to study glucose transport and antidiabetic mechanisms. It demonstrates Kis of 97.4 nM (human SGLT1) and 43.5 nM (rat SGLT1) and is supplied with high purity for biochemical and cellular assays.

  • Selective SGLT1 inhibitor with nanomolar potency.
  • Reported Ki: 97.4 nM (human), 43.5 nM (rat).
  • High purity suitable for research (98.8%).
  • Molecular formula C26H40N4O8; molecular weight 536.62 g/mol.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-36806 Supplier Medchemexpress LLC Supplier No. HY12379710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KGA-2727 is a selective, high-affinity, orally active SGLT1 inhibitor used in preclinical research to study glucose transport and antidiabetic mechanisms. It demonstrates Kis of 97.4 nM (human SGLT1) and 43.5 nM (rat SGLT1) and is supplied with high purity for biochemical and cellular assays.

  • Selective SGLT1 inhibitor with nanomolar potency.
  • Reported Ki: 97.4 nM (human), 43.5 nM (rat).
  • High purity suitable for research (98.8%).
  • Molecular formula C26H40N4O8; molecular weight 536.62 g/mol.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.