Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',4,4',5-Pentachlorobiphenyl Solution, 100μg/mL in Hexane, Ultra Scientific

Catalog No. USRPC102S
Encompass
Change view
Click to view available options
Quantity:
1 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RPC-102S 1 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-RPC-102S Supplier Agilent Technologies Supplier No. RPC102S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57465-28-8 , 110-54-3
Molecular Formula C12H5Cl5
Molecular Weight (g/mol) 326.422
InChI Key REHONNLQRWTIFF-UHFFFAOYSA-N
PubChem CID 63090
ChEBI CHEBI:34317
IUPAC Name 1,2,3-trichloro-5-(3,4-dichlorophenyl)benzene
SMILES C1=CC(=C(C=C1C2=CC(=C(C(=C2)Cl)Cl)Cl)Cl)Cl

Specifications

Physical Form Liquid
Quantity 1 x 2 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 3,3',4,4',5-Pentachlorobiphenyl Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.