Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,4,4-Tetrachlorosulfolane 96.0+%, TCI America™
SDP

Catalog No. T25041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25041G Supplier TCI America Supplier No. T25041G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3737-41-5
Molecular Formula C4H4Cl4O2S
Molecular Weight (g/mol) 257.934
MDL Number MFCD00154882
InChI Key GCAXGCSCRRVVLF-UHFFFAOYSA-N
Synonym 3,3,4,4-Tetrachlorotetrahydrothiophene 1,1-Dioxide
PubChem CID 77328
IUPAC Name 3,3,4,4-tetrachlorothiolane 1,1-dioxide
SMILES C1C(C(CS1(=O)=O)(Cl)Cl)(Cl)Cl

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 257.93
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,4,4-Tetrachlorosulfolane
EINECSNumber (5)-5824
RTECSNumber XN0870000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.