Learn More
Medchemexpress LLC 3-{[(3,4-dichlorophenyl)amino]carbonyl}bicyclo[2.2.1]hept-5-ene-2-carboxylic acid | 199735-88-1 | MFCD00167693 | >98.0% | 326.17 g·mol⁻¹ | C15H13Cl2NO3 | 50 MG
SDP
Catalog No. 5000242454
Encompass_Preferred
CADD522 is a small-molecule inhibitor of RUNX2-DNA binding used in research to downregulate RUNX2-mediated transcription. It has demonstrated antiproliferative and antimetastatic effects in cell-based and animal cancer studies and is supplied as a solid for laboratory use.
  • Inhibits RUNX2-DNA binding with reported IC50 ≈ 10 nM.
  • Demonstrated activity in in vitro and in vivo cancer models.
  • High purity appropriate for biochemical and cell-based assays.
  • Provided in small research quantities for screening and mechanistic studies.
  • Stable solid form; store at -20 °C under recommended conditions.
Catalog No. 50-002-42454 Supplier Medchemexpress LLC Supplier No. HY10799950MG
CADD522 is a small-molecule inhibitor of RUNX2-DNA binding used in research to downregulate RUNX2-mediated transcription. It has demonstrated antiproliferative and antimetastatic effects in cell-based and animal cancer studies and is supplied as a solid for laboratory use.
  • Inhibits RUNX2-DNA binding with reported IC50 ≈ 10 nM.
  • Demonstrated activity in in vitro and in vivo cancer models.
  • High purity appropriate for biochemical and cell-based assays.
  • Provided in small research quantities for screening and mechanistic studies.
  • Stable solid form; store at -20 °C under recommended conditions.