Learn More
Medchemexpress LLC 3-((3,4-dichlorophenyl)carbamoyl)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid | 199735-88-1 | MFCD00167693 | 98.2% | 326.17 g·mol⁻¹ | C15H13Cl2NO3 | 100 MG
SDP
Catalog No. 5000242145
Encompass_Preferred
CADD522 is a small-molecule inhibitor of RUNX2-DNA binding used for laboratory research. It downregulates RUNX2-mediated transcription, exhibits an IC50 of about 10 nM, and has shown inhibition of primary tumor growth and experimental metastasis in preclinical studies. Chemical identifiers include CAS 199735-88-1, formula C15H13Cl2NO3, and molecular weight 326.17 g·mol⁻¹.
  • High purity (98.2%).
  • Potent RUNX2-DNA binding inhibition (IC50 ≈ 10 nM).
  • Suitable for in vitro and preclinical research applications.
  • White to off-white solid, soluble in DMSO as a 10 mM solution.
  • Available as a 100 mg solid package.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-42145 Supplier Medchemexpress LLC Supplier No. HY107999100MG
CADD522 is a small-molecule inhibitor of RUNX2-DNA binding used for laboratory research. It downregulates RUNX2-mediated transcription, exhibits an IC50 of about 10 nM, and has shown inhibition of primary tumor growth and experimental metastasis in preclinical studies. Chemical identifiers include CAS 199735-88-1, formula C15H13Cl2NO3, and molecular weight 326.17 g·mol⁻¹.
  • High purity (98.2%).
  • Potent RUNX2-DNA binding inhibition (IC50 ≈ 10 nM).
  • Suitable for in vitro and preclinical research applications.
  • White to off-white solid, soluble in DMSO as a 10 mM solution.
  • Available as a 100 mg solid package.
  • For research use only; not for human or diagnostic use.