Learn More
Sigma Aldrich 3-(3,4-Diethoxyphenyl)-2-propenoic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 8-hydroxy-2-quinolinecarboxylic acid (8hqc) is a tridentate chelating agent
- It reacts with 2-aminophenol to form a benzoxazole derivative, which can undergo fluorescence quenching in water, making it a viable candidate for developing a probe for detecting water in aprotic organic solvents
- The analysis of mid-IR and raman spectra of 8hqc shows the presence of seven tautomers.8-hydroxy-2-quinolinecarboxylic acid may be used in the preparation of bis (8-hydroxyquinoline-2-carboxylato-+_3n,o,oα) cobalt (ii) trihydrate
Specifications
Specifications
| Molecular Formula | C13H16O4 |
| Linear Formula | C13H16O4 |
| MDL Number | MFCD02256162 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 236.26 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.