Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine Sesquihydrate 98.0+%, TCI America™
SDP

Catalog No. D18175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18175G 5 g
D181725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18175G Supplier TCI America Supplier No. D18175G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 555-30-6
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.217
MDL Number MFCD00004186
InChI Key CJCSPKMFHVPWAR-JTQLQIEISA-N
Synonym methyldopa, alphamethyldopa, aldomet, alpha medopa, dopamet, l-alpha-methyldopa, alpha-methyl dopa, baypresol, hyperpax, presolisin
PubChem CID 38853
ChEBI CHEBI:61058
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid
SMILES CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)N

Specifications

Color White
Quantity 5 g
Formula Weight 211.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine Sesquihydrate
EINECSNumber (9)-0636
RTECSNumber AY5950000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.