Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(3,4-dihydroxyphenyl)-7-hydroxychromen-4-one | 485-63-2 | MFCD00143002 | 99.8% | 270.24 g/mol | C15H10O5 | 10MM 1ML
SDP

Supplier:  Medchemexpress LLC HY12495310MM 1ML

Encompass_Preferred

7,3',4'-Trihydroxyisoflavone is an isoflavone research compound provided as a ready-to-use 10 mM solution in DMSO for biochemical and cell-based studies. It functions as a Cot/MKK4 inhibitor and is used to investigate signaling pathways, enzyme inhibition, and isoflavone metabolism.

  • Ready-to-use 10 mM solution in DMSO, one mL vial.
  • High purity: 99.8% (reported).
  • Chemical formula C15H10O5 and molecular weight 270.24 g/mol.
  • CAS number 485-63-2 for unambiguous identification.
  • Storage recommendations: powder at -20°C; solution at -80°C for long-term stability.

Catalog No. 50-002-35170


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.