Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone 98.0+%, TCI America™
SDP

Catalog No. T15031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15031G Supplier TCI America Supplier No. T15031G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2455-14-3
Molecular Formula C28H40O2
Molecular Weight (g/mol) 408.626
MDL Number MFCD00051798
InChI Key GQIGHOCYKUBBOE-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetra-tert-butyldiphenoquinone, 3,3',5,5'-tetra-tert-butyl-4,4'-diphenoquinone, 3,3',5,5'-tetra-tert-butyl-1,1'-bi cyclohexylidene-2,2',5,5'-tetraene-4,4'-dione, unii-4m92z38q9a, 3,3',5,5'-tetra-tert-butyl-4,4'-dibenzoquinone, bi-2,5-cyclohexadien-1-ylidene-4,4'-dione, 3,3',5,5'-tetra-tert-butyl, 2,2',6,6'-tetra-tert-butyldiphenylquinone, 2,6-ditert-butyl-4-3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene cyclohexa-2,5-dien-1-one, pubchem13798, rarechem bw gc 0005
PubChem CID 225283
IUPAC Name 2,6-ditert-butyl-4-(3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)cyclohexa-2,5-dien-1-one
SMILES CC(C)(C)C1=CC(=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)C=C(C1=O)C(C)(C)C

Specifications

Color Black-Purple
Quantity 1 g
Formula Weight 408.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,5,5′-Tetra-tert-butyl-4,4′-diphenoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.