Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetra-tert-butyl-4,4'-stilbenequinone 95.0+%, TCI America™
SDP

Catalog No. T10675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10675G 5 g
T10671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10675G Supplier TCI America Supplier No. T10675G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 809-73-4
Molecular Formula C30H42O2
MDL Number MFCD00060073

Specifications

Quantity 5 g
Formula Weight 434.66
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,5,5′-Tetra-tert-butyl-4,4′-stilbenequinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.