Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98%, Thermo Scientific Chemicals

Catalog No. AC448300020
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448300020 2 g
AC448300100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448300020 Supplier Thermo Scientific Chemicals Supplier No. 448300020
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2455-14-3
Molecular Formula C28H40O2
Molecular Weight (g/mol) 408.63
InChI Key GQIGHOCYKUBBOE-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetra-tert-butyldiphenoquinone, 3,3',5,5'-tetra-tert-butyl-4,4'-diphenoquinone, 3,3',5,5'-tetra-tert-butyl-1,1'-bi cyclohexylidene-2,2',5,5'-tetraene-4,4'-dione, unii-4m92z38q9a, 3,3',5,5'-tetra-tert-butyl-4,4'-dibenzoquinone, bi-2,5-cyclohexadien-1-ylidene-4,4'-dione, 3,3',5,5'-tetra-tert-butyl, 2,2',6,6'-tetra-tert-butyldiphenylquinone, 2,6-ditert-butyl-4-3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene cyclohexa-2,5-dien-1-one, pubchem13798, rarechem bw gc 0005
PubChem CID 225283
IUPAC Name 2,6-ditert-butyl-4-(3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)cyclohexa-2,5-dien-1-one
SMILES CC(C)(C)C1=CC(=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)C=C(C1=O)C(C)(C)C

Specifications

Melting Point 247.0°C to 248.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 2 g
Formula Weight 408.63
Percent Purity 98%
Chemical Name or Material 3,3',5,5'-Tetra-tert-butyldiphenoquinone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 219-527-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.