Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98+%

Catalog No. AAL0774304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0774304 2 g
AAL0774309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0774304 Supplier Thermo Scientific Chemicals Supplier No. L0774304
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2455-14-3 | C28H40O2 | 408.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2455-14-3
Molecular Formula C28H40O2
Molecular Weight (g/mol) 408.626
MDL Number MFCD00051798
InChI Key GQIGHOCYKUBBOE-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetra-tert-butyldiphenoquinone, 3,3',5,5'-tetra-tert-butyl-4,4'-diphenoquinone, 3,3',5,5'-tetra-tert-butyl-1,1'-bi cyclohexylidene-2,2',5,5'-tetraene-4,4'-dione, unii-4m92z38q9a, 3,3',5,5'-tetra-tert-butyl-4,4'-dibenzoquinone, bi-2,5-cyclohexadien-1-ylidene-4,4'-dione, 3,3',5,5'-tetra-tert-butyl, 2,2',6,6'-tetra-tert-butyldiphenylquinone, 2,6-ditert-butyl-4-3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene cyclohexa-2,5-dien-1-one, pubchem13798, rarechem bw gc 0005
PubChem CID 225283
IUPAC Name 2,6-ditert-butyl-4-(3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)cyclohexa-2,5-dien-1-one
SMILES CC(C)(C)C1=CC(=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)C=C(C1=O)C(C)(C)C

Specifications

Melting Point 242°C to 246°C
Quantity 2 g
Beilstein 1916759
Formula Weight 408.63
Percent Purity ≥98%
Chemical Name or Material 3,3',5,5'-Tetra-tert-butyldiphenoquinone
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 219-527-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.