Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3',3,5',5-Tetrabromophenolphthalein ethyl ester

Catalog No. 867781G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-778-1G 1 g
86-778-250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-778-1G Supplier Sigma Aldrich Supplier No. 867781G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3α,3α,5α,5α-tetrabromophenolphthalein ethyl ester (tbpe) is an anionic dye used as a counter ion in the spectrophotometric determination of cationic surfactants
  • Tbpe is also used in the spectrometric determination of aliphatic primary, secondary and tertiary amines

Specifications

CAS 1176-74-5
Molecular Formula C22H14Br4O4
Linear Formula C22H14Br4O4
MDL Number MFCD00066387
Quantity 1 g
Synonym TBPE
Molecular Weight (g/mol) 661.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.