Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol, 97%, Thermo Scientific™

Catalog No. AAB2055314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2055314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2055314 Supplier Thermo Scientific Chemicals Supplier No. B2055314

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87901-76-6
Molecular Formula C17H8F12O
Molecular Weight (g/mol) 456.23
MDL Number MFCD00042475
InChI Key DUCQGUCBWHWEBT-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetrakis trifluoromethyl benzhydrol, bis 3,5-bis trifluoromethyl phenyl methanol, maybridge1_002893, di 3,5-di trifluoromethyl phenyl methanol, 3,3,5,5-tetrakis trifluoromethyl benzhydrol, benzenemethanol, a-3,5-bis trifluoromethyl phenyl-3,5-bis trifluoromethyl
PubChem CID 2776818
IUPAC Name bis[3,5-bis(trifluoromethyl)phenyl]methanol
SMILES OC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 147°C to 149°C
Quantity 25 g
Formula Weight 456.23
Percent Purity 97%
Chemical Name or Material 3,3',5,5'-Tetrakis(trifluoromethyl)benzhydrol
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.