Learn More
Sigma Aldrich Fine Chemicals Biosciences 3,3′,5,5′-Tetramethylbenzidine Liquid Substrate, Slow Kinetic Form, for ELISA | 100 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T4319100ML
Encompass
3,3′,5,5′-Tetramethylbenzidine Liquid Substrate, Slow Kinetic Form, for ELISA has been used as a substrate for anti-bovine IgG-peroxidase conjugated antibody and for horse radish peroxidase-conjugated mouse anti-mouse IgG in Poly (ADP-ribose) binding assays.
Catalog No. NC1709094