Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetramethylbenzidine dihydrochloride, 98%

Catalog No. AAJ6033206
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6033206 5 g
AAJ6033203 1 g
AAJ6033288 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6033206 Supplier Thermo Scientific Chemicals Supplier No. J6033206
Add to Cart

CAS: 207738-08-7 | C16H22Cl2N2 | 313.27 g/mol

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

Solubility
Soluble in DMSO (∼1.7 mg/ml), and 1:300 DMSO:PBS (pH 7.2) (0.03 mg/ml).

Notes
Moisture and light sensitive. Store away from oxidizing agents, heat, light, acids, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207738-08-7
Molecular Formula C16H22Cl2N2
Molecular Weight (g/mol) 313.27
MDL Number MFCD00150104
InChI Key NYNRGZULARUZCC-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride
PubChem CID 19836601
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride
SMILES Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1

Specifications

Color White
Quantity 5 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in DMSO (∽1.7mg/ml),and 1:300 DMSO:PBS (pH 7.2) (0.03mg/ml).
Formula Weight 313.27
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
Hazard Category H302+H312+H332-H341
Hazard Statement GHS H Statement
H341-H302-H315-H319
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.